Learn More
FADD Monoclonal Antibody (OTI1C11)

Description
FADD Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | FADD |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI1C11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FADD |
| Gene Accession No. | Q13158 |
| Gene Alias | Fadd; Fas (TNFRSF6)-associated via death domain; Fas associated via death domain; FAS-associated death domain protein; FAS-associating death domain-containing protein; Fas-associating protein with death domain; GIG3; Growth-inhibiting gene 3 protein; mediator of receptor induced toxicity; mediator of receptor-induced toxicity; MGC8528; MORT1; Mort1/FADD; protein FADD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.