Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FADS1 Recombinant Rabbit Monoclonal Antibody (1M8L2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542587
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, Mouse liver, Rat lung, Rat liver, Rat brain. Immunogen sequence: RRHPGGSRVI SHYAGQDATD PFVAFHINKG LVKKYMNSLL IGELSPEQPS FEPTKNKELT DEFRELRATV ERMGLMKANH VFFLLYLLHI LLLDGAAWLT L.
The protein encoded by this gene is a member of the fatty acid desaturase gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13. 1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization.
Specifications
FADS1 | |
Recombinant Monoclonal | |
0.4 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60427, Q920L1, Q920R3 | |
FADS1 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human FADS1 (O60427). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
1M8L2 | |
Unconjugated | |
FADS1 | |
0710001O03Rik; A930006B21Rik; Acyl-CoA (8-3)-desaturase; AI317215; BC269730_2; D5D; delta(5) desaturase; delta(5) fatty acid desaturase; delta-5 desaturase; delta-5 fatty acid desaturase; DSD; FADS1; FADS6; FADSD5; Fatty acid desaturase 1; FLJ38956; FLJ90273; linoleoyl-CoA desaturase (delta-6-desaturase)-like 1; LLCDL1; TU12 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3992, 76267, 84575 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction