Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FADS1 Recombinant Rabbit Monoclonal Antibody (JE55-63)
GREENER_CHOICE

Catalog No. PIMA542500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542500 Supplier Invitrogen™ Supplier No. MA542500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: Rat liver tissue lysate, mouse liver tissue lysate, mouse brain tissue lysate, rat lung tissue lysate, Hela cell lysate, A549 cell lysate, Hela, PANC-1, human liver carcinoma tissue, human thyroid tissue, human colon carcinoma tissue, human breast carcinoma tissue, human stomach carcinoma tissue, human small intestine tissue, A549. Subcellular Location: Mitochondrion, Endoplasmic reticulum membrane.

The protein encoded by this gene is a member of the fatty acid desaturase gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13. 1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FADS1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE55-63
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene FADS1
Gene Accession No. O60427, Q920L1, Q920R3
Gene Alias 0710001O03Rik; A930006B21Rik; Acyl-CoA (8-3)-desaturase; AI317215; BC269730_2; D5D; delta(5) desaturase; delta(5) fatty acid desaturase; delta-5 desaturase; delta-5 fatty acid desaturase; DSD; FADS1; FADS6; FADSD5; Fatty acid desaturase 1; FLJ38956; FLJ90273; linoleoyl-CoA desaturase (delta-6-desaturase)-like 1; LLCDL1; TU12
Gene Symbols FADS1
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal Human FADS1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3992, 76267, 84575
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.