Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FADS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119108
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119108 200 μL
PIPA5122007 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119108 Supplier Invitrogen™ Supplier No. PA5119108
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FADS2 Polyclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is a member of the fatty acid desaturase (FADS) gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13. 1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FADS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Fads2
Gene Accession No. O95864
Gene Alias 2900042M13Rik; Acyl-CoA 6-desaturase; D6D; delta(6) desaturase; delta(6) fatty acid desaturase; delta-6 desaturase; delta-6 fatty acid desaturase; delta-6-desaturase; DES6; FADS2; Fadsd2; FADSD6; Fatty acid desaturase 2; linoleoyl-CoA desaturase (delta-6-desaturase)-like 2; LLCDL2; SLL0262; TU13
Gene Symbols Fads2
Host Species Rabbit
Immunogen Recombinant FADS2 (Met1-Val131) expressed in E.coli.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 444997, 9415
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.