Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FADS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560956
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560956 Supplier Invitrogen™ Supplier No. PA560956
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLPTFCWEQI RAHDQPGDKW LVIERRVYDI SRWAQRHPGG SRLIGHHGAE D Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

The protein encoded by this gene is a member of the fatty acid desaturase (FADS) gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13.1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FADS3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FADS3
Gene Accession No. Q9Y5Q0
Gene Alias AI464531; CYB5RP; cytochrome b5-related protein; delta(13) desaturase; Delta(13) fatty acid desaturase; delta-9 fatty acid desaturase; delta-9-desaturase; FADS3; Fatty acid desaturase 3; fatty acid desaturase 3; LOW QUALITY PROTEIN: fatty acid desaturase 3; linoleoyl-CoA desaturase (delta-9-desaturase)-like 3; LLCDL3; MNCb-0629; putative fatty acid desaturase
Gene Symbols FADS3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FADS3. Recombinant protein control fragment (Product #RP-100011).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3995
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.