Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FADS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571087 Supplier Invitrogen™ Supplier No. PA571087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 86%; Rat: 86%.

FADS6 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; lipid metabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FADS6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FADS6
Gene Accession No. Q8N9I5
Gene Alias BC044804; FADS6; Fatty acid desaturase 6; fatty acid desaturase domain family, member 6; FP18279; OTTMUSG00000021749; RGD1311950
Gene Symbols FADS6
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human FADS6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283985
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.