Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FADS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553491 Supplier Invitrogen™ Supplier No. PA553491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GHSIISCHVE HHLFPRLSDN MCLKVKPVVS QFLREKQLPY NEDSYLARFQ LFLRRYEEFM VQAPPITELV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.

FADS6 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; lipid metabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FADS6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FADS6
Gene Accession No. Q8N9I5
Gene Alias BC044804; FADS6; Fatty acid desaturase 6; fatty acid desaturase domain family, member 6; FP18279; OTTMUSG00000021749; RGD1311950
Gene Symbols FADS6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FADS6. Recombinant protein control fragment (Product #RP-91232).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.