Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAF1 Monoclonal Antibody (OTI10G6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA804153

Description
Interaction of Fas ligand (TNFSF6) with the FAS antigen (TNFRSF6) mediates programmed cell death, also called apoptosis, in a number of organ systems. The protein encoded by this gene binds to FAS antigen and can initiate apoptosis or enhance apoptosis initiated through FAS antigen. Initiation of apoptosis by the protein encoded by this gene requires a ubiquitin-like domain but not the FAS-binding domain.Specifications
FAF1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UNN5 | |
FAF1 | |
Human recombit protein fragment corresponding to amino acids 415-650 of human FAF1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin) | |
OTI10G6 | |
Unconjugated | |
FAF1 | |
CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
11124 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction