Learn More
Invitrogen™ FAF1 Polyclonal Antibody

Description
A suggested positive control is Jurkat cell lysate. PA5-20314 can be used with blocking peptide PEP-0434.
Specifications
Specifications
| Antigen | FAF1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | FAF1 |
| Gene Accession No. | P54731, Q924K2, Q9UNN5 |
| Gene Alias | AA408698; CGI-03; Dffrx; Faf1; Fam; Fas (TNFRSF6) associated factor 1; Fas associated factor 1; Fas-associated factor 1; hFAF1; HFAF1s; TNFRSF6-associated factor 1; UBX domain protein 3A; UBX domain-containing protein 12; UBX domain-containing protein 3A; UBXD12; UBXN3A |
| Gene Symbols | FAF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.