Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAF2 Mouse anti-Human, Clone: 3H6D2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | FAF2 |
|---|---|
| Clone | 3H6D2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7047
![]()
|
Proteintech Group Inc
666291IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.Specifications
| FAF2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ETEA, FAF2, FAS associated factor 2, KIAA0887, Protein ETEA, UBXD8, UBXN3B | |
| FAF2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3H6D2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96CS3 | |
| 23197 | |
| FAF2 Fusion Protein Ag21536 | |
| Primary | |
| -20°C | |
| FAF2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title