Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAH Recombinant Rabbit Monoclonal Antibody (HL1970)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547242
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT).
Specifications
FAH | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P16930, P25093, P35505 | |
FAH | |
Recombinant fragment of human FAH. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
HL1970 | |
Unconjugated | |
FAH | |
Beta-diketonase; FAA; FAH; fb59b12; fumarylacetoacetase; Fumarylacetoacetate hydrolase; fumarylacetoacetate hydrolase (fumarylacetoacetase); LOW QUALITY PROTEIN: fumarylacetoacetase; wu:fb59b12; zgc:55316 | |
Rabbit | |
Protein A | |
RUO | |
14085, 2184, 29383 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction