Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542050 Supplier Invitrogen™ Supplier No. PA542050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AATICKSNFK YMYWTMLQQL THHSVNGCNL RPGDLLASGT ISGPEPENFG Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.

This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAH
Gene Accession No. P16930, P35505
Gene Alias Beta-diketonase; FAA; FAH; fb59b12; fumarylacetoacetase; Fumarylacetoacetate hydrolase; fumarylacetoacetate hydrolase (fumarylacetoacetase); LOW QUALITY PROTEIN: fumarylacetoacetase; wu:fb59b12; zgc:55316
Gene Symbols FAH
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FAH (aa 311-360).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14085, 2184
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.