Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583705
Description
Immunogen sequence: ACKLLDMELE MAFFVGPGNR LGEPIPISKA HEHIFGMVLM NDWSARDIQK WEYVPLGPFL GKSFGTTVSP WVVPMDALMP FAVPNPKQDP RPLPYLCHDE PYT.
This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT).
Specifications
FAH | |
Polyclonal | |
Unconjugated | |
FAH | |
Beta-diketonase; FAA; FAH; fb59b12; fumarylacetoacetase; Fumarylacetoacetate hydrolase; fumarylacetoacetate hydrolase (fumarylacetoacetase); LOW QUALITY PROTEIN: fumarylacetoacetase; wu:fb59b12; zgc:55316 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2184 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P16930 | |
FAH | |
Recombinant protein corresponding to Human FAH. Recombinant protein control fragment (Product #RP-98043). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction