Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583551 Supplier Invitrogen™ Supplier No. PA583551
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INLSVNLKGE GMSQAATICK SNFKYMYWTM LQQLTHHSVN GCNLRPGDLL ASGTISGPEP ENFGSMLELS WKGTKPIDLG NGQTRKFLLD GD Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 88%.

This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAH
Gene Accession No. P16930
Gene Alias Beta-diketonase; FAA; FAH; fb59b12; fumarylacetoacetase; Fumarylacetoacetate hydrolase; fumarylacetoacetate hydrolase (fumarylacetoacetase); LOW QUALITY PROTEIN: fumarylacetoacetase; wu:fb59b12; zgc:55316
Gene Symbols FAH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAH. Recombinant protein control fragment (Product #RP-98042).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.