Learn More
FAH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA530027
Description
Recommended positive controls: A549, mouse liver, mouse kidney. Predicted reactivity: Mouse (88%), Rat (88%), Chicken (81%), Rhesus Monkey (95%), Chimpanzee (98%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT).
Specifications
FAH | |
Polyclonal | |
Unconjugated | |
FAH | |
Beta-diketonase; FAA; FAH; fb59b12; fumarylacetoacetase; Fumarylacetoacetate hydrolase; fumarylacetoacetate hydrolase (fumarylacetoacetase); LOW QUALITY PROTEIN: fumarylacetoacetase; wu:fb59b12; zgc:55316 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14085, 2184 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.38 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P16930, P35505 | |
FAH | |
Recombinant fragment corresponding to a region within amino acids 185 and 419 of Human Fumarylacetoacetase | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.