Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAHD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570890
Description
This target displays homology in the following species: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rat: 100%; Yeast: 86%; Zebrafish: 93%.
Probable mitochondrial acylpyruvase which is able to hydrolyze acetylpyruvate and fumarylpyruvate in vitro (PubMed:15551868, PubMed:21878618). Also has oxaloacetate decarboxylase activity (PubMed:25575590). [UniProt].
Specifications
FAHD1 | |
Polyclonal | |
Unconjugated | |
FAHD1 | |
1110025H10Rik; Acylpyruvase FAHD1, mitochondrial; acylpyruvate hydrolase; AV002085; C16orf36; FAH domain-containing protein 1; Fahd1; fumarylacetoacetate hydrolase domain containing 1; fumarylacetoacetate hydrolase domain-containing protein 1; hypothetical protein LOC553762; MNCb-4134; OAA decarboxylase; oxaloacetate decarboxylase; RGD1304560; Unknown (protein for MGC:138003); wu:fj09c02; YISK like/RJD15; YISKL; yisK-like protein; zgc:109929 | |
Rabbit | |
Affinity chromatography | |
RUO | |
81889 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6P587 | |
FAHD1 | |
synthetic peptide directed towards the middle region of human FAHD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction