Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565362
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565362 Supplier Invitrogen™ Supplier No. PA565362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DECKKKGLPWT LAKSFTASCP VSAFVPKEKI PDPHKLKLWL KVNGELRQEG ETSSMIFSIP YIISYVSKII TLEEGDIILT GT Highest antigen sequence identity to the following orthologs - mouse 93%, rat 94%.

Probable mitochondrial acylpyruvase which is able to hydrolyze acetylpyruvate and fumarylpyruvate in vitro (PubMed:15551868, PubMed:21878618). Also has oxaloacetate decarboxylase activity (PubMed:25575590). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAHD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAHD1
Gene Accession No. Q6P587
Gene Alias 1110025H10Rik; Acylpyruvase FAHD1, mitochondrial; acylpyruvate hydrolase; AV002085; C16orf36; FAH domain-containing protein 1; Fahd1; fumarylacetoacetate hydrolase domain containing 1; fumarylacetoacetate hydrolase domain-containing protein 1; hypothetical protein LOC553762; MNCb-4134; OAA decarboxylase; oxaloacetate decarboxylase; RGD1304560; Unknown (protein for MGC:138003); wu:fj09c02; YISK like/RJD15; YISKL; yisK-like protein; zgc:109929
Gene Symbols FAHD1
Host Species Rabbit
Immunogen Recombinant Human FAHD1. Recombinant protein control fragment (Product #RP-104773).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81889
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.