Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAHD1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982639
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2639 Supplier Bioss Supplier No. BS13132R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRF2 is a ubiquitously expressed protein that is implicated in the control of telomere length. TRF2, like TRF1 contains a Myb-related DNA binding motif. It binds to duplex TTAGGG repeats and is localized to all human telomeres in metaphase chromosomes. TRF2 is thought to protect chromosome ends by maintaining the correct structure at telomere termini. The use of mutant forms of TRF2 has implicated a role for TRF2 in the prevention of senescence in primary human cells. Recently, it has been shown that inhibition of TRF2 resulted in apoptosis in a subset of mammalian cell types.

Specifications

Antigen FAHD1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAHD1
Gene Accession No. Q6P587
Gene Alias 1110025H10Rik; Acylpyruvase FAHD1, mitochondrial; acylpyruvate hydrolase; AV002085; BOS_22457; C16orf36; FAH domain-containing protein 1; Fahd1; fumarylacetoacetate hydrolase domain containing 1; Fumarylacetoacetate hydrolase domain-containing protein 1; hypothetical protein LOC553762; MNCb-4134; OAA decarboxylase; Oxaloacetate decarboxylase; RGD1304560; wu:fj09c02; YISK like/RJD15; YISKL; YisK-like protein; zgc:109929
Gene Symbols FAHD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAHD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81889
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.