Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAHD2A Antibody (OTI6D9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271886FR
Description
FAHD2A Monoclonal specifically detects FAHD2A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| FAHD2A | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| FAHD2A | |
| Full length recombinant protein of human FAHD2A (NP_057128) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6D9 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CGI-105, EC 3.-, fumarylacetoacetate hydrolase domain containing 1, fumarylacetoacetate hydrolase domain containing 2A, fumarylacetoacetate hydrolase domain-containing protein 2A, MGC131995 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 51011.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction