Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAHD2A Monoclonal Antibody (OTI6D9)
GREENER_CHOICE

Catalog No. PIMA525048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525048 Supplier Invitrogen™ Supplier No. MA525048
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAHD2A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

May have hydrolase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAHD2A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI6D9
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FAHD2A
Gene Accession No. Q96GK7
Gene Alias 1500003K14Rik; B430104H02Rik; CGI-105; CGI-105 protein; Fahd2; FAHD2A; fumarylacetoacetate hydrolase domain containing 1; fumarylacetoacetate hydrolase domain containing 2A; fumarylacetoacetate hydrolase domain-containing protein 2; Fumarylacetoacetate hydrolase domain-containing protein 2A; LOC100062565; LOC100731203; RGD1563674
Gene Symbols FAHD2A
Host Species Mouse
Immunogen Full length human recombinant protein of FAHD2A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51011
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.