Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAHD2A Monoclonal Antibody (OTI6D9), TrueMAB™, OriGene
SDP

Catalog No. 502542670
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2670 30 μL
50-167-2079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2670 Supplier OriGene Supplier No. TA500939S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAHD2A Monoclonal antibody specifically detects FAHD2A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen FAHD2A
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6D9
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FAHD2A
Gene Accession No. Q96GK7
Gene Alias 1500003K14Rik; B430104H02Rik; CGI-105; CGI-105 protein; Fahd2; FAHD2A; fumarylacetoacetate hydrolase domain containing 1; fumarylacetoacetate hydrolase domain containing 2A; fumarylacetoacetate hydrolase domain-containing protein 2; Fumarylacetoacetate hydrolase domain-containing protein 2A; LOC100062565; LOC100731203; RGD1563674
Host Species Mouse
Immunogen Full length human recombinant protein of human FAHD2A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51011
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.