Learn More
Invitrogen™ FAIM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587476
Description
Immunogen sequence: MTDLVAVWDV ALSDGVHKIE FEHGTTSGKR VVYVDGKEEI RKEWMFKLVG KETFYVGAAK TKATINIDAI SGFAYEYTLE INGKSLKKYM EDRSKTTNTW VLHMDGENFR IVLEKDAMDV WCNGKKLETA GEFVDDGTET HFSIGNHDCY IKAVSSGKRK EGIIHTLIVD NREIPEIAS; Positive Samples: U-87MG, BT-474; Cellular Location: Cytoplasm.
Plays a role as an inducible effector molecule that mediates Fas resistance produced by surface Ig engagement in B cells.
Specifications
FAIM | |
Polyclonal | |
Unconjugated | |
FAIM | |
Faim; FAIM 1; FAIM1; Fas apoptotic inhibitory molecule; fas apoptotic inhibitory molecule 1; rFAIM | |
Rabbit | |
Affinity chromatography | |
RUO | |
55179 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.77 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NVQ4 | |
FAIM | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-179 of human FAIM (NP_0606171). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.