Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAIM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575639
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Plays a role as an inducible effector molecule that mediates Fas resistance produced by surface Ig engagement in B cells.
Specifications
FAIM | |
Polyclonal | |
Unconjugated | |
FAIM | |
Faim; FAIM 1; FAIM1; Fas apoptotic inhibitory molecule; fas apoptotic inhibitory molecule 1; rFAIM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55179 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NVQ4 | |
FAIM | |
Recombinant full length Human FAIM. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction