Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAIM3 Monoclonal Antibody (OTI1E6)
GREENER_CHOICE

Catalog No. PIMA526353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526353 Supplier Invitrogen™ Supplier No. MA526353
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAIM3 Monoclonal Antibody for IHC (P)

Apoptosis is an important process by which normal tissue homeostasis and function are maintained. One of the major signals that regulate this process is mediated by the activation of the Fas receptor by its ligand. This leads to the formation of a Fas-associated death domain (FADD)- containing death-inducing signaling complex and the activation of caspase-8, which in turn activates downstream effector caspases, such as caspase-3 and -7. Recent experiments have shown that overexpression of Toso, a novel regulator of Fas-induced apoptosis in lymphoid cells, in Jurkat cells as well as transgenic mice render these cells resistant to Fas-induced apoptosis but not to TRAIL-induced apoptosis. Furthermore, Toso was found to associate with FADD, suggesting that Toso functions by disrupting the formation of the death-inducing signaling complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAIM3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Fcmr
Gene Accession No. O60667
Gene Alias 1810037B05Rik; Faim3; Fas apoptotic inhibitory molecule 3; Fc fragment of IgM receptor; Fc mu receptor; FCMR; FcmuR; IgM Fc fragment receptor; IgM Fc receptor; immunoglobulin mu Fc receptor; regulator of Fas-induced apoptosis Toso; TOSO
Gene Symbols Fcmr
Host Species Mouse
Immunogen Full length human recombinant protein of FAIM3 produced in HEK293 Cells.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9214
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.