Learn More
Promega FAK Kinase Enzyme System, 1mg
Catalog No. PRV1972 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV1972 1 mg
1 options
Catalog No. PRV1972 Supplier Promega Supplier No. V1972
Add to Cart
Add to Cart
Request a Quote
Recombinant human FAK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. FAK is involved in signal transduction from integrin-enriched focal adhesion sites that mediate cell contact with the extracellular matrix.
  • Easily Screen and Profile FAK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C