Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAK, p Tyr407 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313085
This item is not returnable.
View return policy
Description
FAK Polyclonal antibody specifically detects FAK in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| FAK | |
| Polyclonal | |
| PBS, 1% BSA, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| EC 2.7.10, FADK 1, FADKFAK-related non-kinase polypeptide, FAK1FRNK, FAKEC 2.7.10.2, focal adhesion kinase 1, pp125FAK, Protein-tyrosine kinase 2, PTK2 protein tyrosine kinase 2 | |
| Carrier-protein conjugated synthetic peptide corresponding to residues around human FAK (phospho Tyr407). The exact sequence is proprietary. | |
| 100 μg | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 5747 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction