Learn More
Falcon™ Cell Culture Multi Flasks
BIOPHARMAGSA_VAGREENER_CHOICE
Catalog No. p-4254206 Shop All Corning Products
Click to view available options
Culture Area:
525 cm2
875 cm2
Surface Coating:
3 Layer TC-Treated
TC-Treated
This item is not returnable. View return policy
This item is not returnable. View return policy
Grow cells faster and easier, increasing productivity in cell culture workflow. Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.
Increase productivity by enabling to grow more cells faster and easier. 3- or 5-layer formats provide 525 cm2 and 875 cm2 cell growth surface area, respectively.
  • Even distribution of media across all layers for homogeneous cell growth
  • Ability to mix cells and reagents in the Falcon Multi-Flask saves time
  • Flexible design allows to pour or aspirate/recover cells using a pipet
  • Consistent surface treatment for predictable scale-up
  • Lot number printed on each flask for traceability
  • Manufactured in compliance to cGMP standards
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Neck Style Straight
Shape Rectangular
Surface Coating 3 Layer TC-Treated
Sterility Sterile
Closure Type Vented
Culture Area 525 cm2
WARNING: Cancer - www.P65Warnings.ca.gov
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.