Learn More
Falcon™ Polystyrene Aspirating Pipets, Without Graduations
BIOPHARMAGREENER_CHOICE
Catalog No. p-162770
Click to view available options
Volume (Metric):
2 mL
5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Manufactured from polystyrene as a safer alternative to glass Pasteur pipets. Falcon™ Disposable Aspirating Pipets are ideal for tissue culture usage or any non-toxic solution that requires aliquoted or aspirating/dispensing in a sterile environment.
  • Sterile, polystyrene, non-plugged, non-graduated pipets
  • Safer alternative to glass Pasteur pipets
  • Can be used for all vacuum-aspirating procedures
  • Noncytotoxic, nonpyrogenic tested to less than 0.1 EU/mL
  • Individually packaged in thermoformed paper-plastic wrap to ensure sterile presentation
Order Info
All sizes come individually wrapped, 200 per case
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Class I Medical
Description Aspirating
Material Polystyrene
Sterility Sterile
For Use With (Application) For tissue culture usage or any non-toxic solution that requires aliquot or aspirating, dispensing in a sterile environment
Volume (Metric) 2 mL
Type Aspirating Pipet
Disposable Single-use
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.