Learn More
Falcon™ Tissue Culture Treated Flasks
Manufactured from optically clear virgin polystyrene for easy viewing of sample or cells. Falcon™ Tissue Culture Treated Flasks features vacuum-gas plasma tissue culture treatment to ensure consistent growth surface.
$286.50 - $559.00
Specifications
Closure Material | Polyethylene |
---|---|
Material | Polystyrene |
For Use With (Application) | For enhanced cell attachment and growth |
Shape | Rectangular |
Surface Coating | TC-Treated |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Type | Neck Style | Culture Area | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Type | Neck Style | Culture Area | Price | Quantity & Availability | ||
08-772-1E
|
Falcon™
353018 |
25 mL | Plug Seal | Canted | 12.5 cm2 |
Case of 100 for $295.00
|
|
||
08-772-1F
|
Falcon™
353107 |
25 mL | Vented | Canted | 12.5 cm2 |
Case of 100 for $373.50
|
|
||
10-126-1C
|
Falcon™
353014 |
50 mL | Plug Seal | Canted | 25 cm2 |
Case of 200 for $509.50
|
|
||
10-126-9
|
Falcon™
353108 |
50 mL | Vented | Canted | 25 cm2 |
Case of 100 for $286.50
|
|
||
08-772-19
|
Falcon™
353082 |
70 mL | Plug Seal | Canted | 25 cm2 |
Case of 200 for $507.50
|
|
||
10-126-10
|
Falcon™
353109 |
70 mL | Vented | Canted | 25 cm2 |
Case of 100 for $300.00
|
|
||
13-680-59
|
Falcon™
353135 |
250 mL | Plug Seal | Canted | 75 cm2 |
Case of 60 for $292.50
|
|
||
10-126-5
|
Falcon™
353024 |
250 mL | Plug Seal | Straight | 75 cm2 |
Case of 100 for $514.50
|
|
||
13-680-65
|
Falcon™
353136 |
250 mL | Vented | Canted | 75 cm2 |
Case of 60 for $330.00
|
|
||
10-126-11
|
Falcon™
353110 |
250 mL | Vented | Straight | 75 cm2 |
Case of 100 for $559.00
|
|
||
08-772-47
|
Falcon™
355000 |
600 mL | Plug Seal | Canted | 150 cm2 |
Case of 40 for $345.50
|
|
||
08-772-48
|
Falcon™
355001 |
600 mL | Vented | Canted | 150 cm2 |
Case of 40 for $377.50
|
|
||
10-126-8
|
Falcon™
353028 |
750 mL | Plug Seal | Straight | 175 cm2 |
Case of 40 for $358.00
|
|
||
10-126-13
|
Falcon™
353112 |
750 mL | Vented | Straight | 175 cm2 |
Case of 40 for $372.50
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
14-826-81
|
Falcon™
353139 |
800 mL | Plug Seal | Canted | 225 cm2 |
Case of 30 for $343.50
|
|
||
14-826-80
|
Falcon™
353138 |
800 mL | Vented | Canted | 225 cm2 |
Case of 30 for $392.00
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Secure stacking provides stability from accidental tips or spills
- Nonpyrogenic tested to less than 0.1 EU/ mL
- Volumetric graduations and writing patch
- Vented caps incorporate a 0.2 μm hydrophobic membrane for optimal gas exchange
- Growth area and volume are nominal
Specifications
Polyethylene | |
For enhanced cell attachment and growth | |
TC-Treated | |
Rectangular |
Polystyrene | |
Rectangular | |
Sterile |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.