Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FALDH Antibody (OTI2A7), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270159UV
Description
FALDH Monoclonal specifically detects FALDH in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
FALDH | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ALDH3A2 | |
Full length recombinant protein of human FALDH (NP_001026976) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI2A7 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Aldehyde dehydrogenase 10, aldehyde dehydrogenase 3 family, member A2, Aldehyde dehydrogenase family 3 member A2, ALDH10FLJ20851, EC 1.2.1, EC 1.2.1.3, FALDHDKFZp686E23276, fatty aldehyde dehydrogenase, Microsomal aldehyde dehydrogenase, SLS | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
224.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction