Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FALZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583905 Supplier Invitrogen™ Supplier No. PA583905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTIASTGQTF QITGNPVTMA GKVITKLPLP ANSKIVAVNV PATQGGIVQV HQ Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 90%.

BPTF (bromodomain and PHD domain transcription factor) is the largest subunit of the ATP-dependent chromatin-remodelling complex, NURF (nucleosome remodelling factor). NURF catalyses ATP-dependent nucleosome sliding and facilitates transcription. BPTF recognises histone H3 tails that are tri-methylated at K4, which marks the transcriptional start site of the vast majority of transcriptionally active genes. BPTF also exhibits some binding to H3 di-methylated at K4. BPTF plays a key role in the development of early mouse embryos, possibly through regulation of the Smad pathway of transcription factors. While BPTF is expressed in low levels in the adult brain and spinal cord, it is expressed in higher levels in the brain in neurodegenerative diseases. It is present in a subset of amyloid-containing plaques in the brains of patients suffering from Alzheimer's disease. Abundantly expressed in the fetal brain. Present throughout the gray and white matter of the developing spinal cord at 18-22 gestational weeks. Expressed at low levels in adult brain and spinal cord and reexpressed in neurodegenerative diseases (at protein level).Tissue specificity: Ubiquitously expressed, with highest levels in testis. Present in kidney, liver and brain. In the brain, highest levels are found in motor cortex (at protein level).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FALZ
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BPTF
Gene Accession No. Q12830
Gene Alias 9430093H17Rik; BPTF; bromodomain and PHD domain transcription factor; bromodomain and PHD finger-containing transcription factor; bromodomain PHD finger transcription factor; FAC1; FALZ; fetal Alz-50 clone 1 protein; fetal Alz-50 reactive clone 1; fetal Alzheimer antigen; nucleosome remodeling factor, large subunit; nucleosome-remodeling factor subunit BPTF; NURF301
Gene Symbols BPTF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BPTF. Recombinant protein control fragment (Product #RP-106184).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2186
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.