Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM101A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566221 Supplier Invitrogen™ Supplier No. PA566221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FFGESIKVNPE PTHEIRCNSE VKYASEKHFQ DKVFYAPVPT ITAYSETIVA APNCTWRNYR SQLTLEPRPR ALRFRSTTII FPKHARSTFR TTLHCSLGRP SRWFTASVQL QLCQDP Highest antigen sequence identity to the following orthologs - mouse 79%, rat 79%.

Involved in the regulation of the perinuclear actin network and nuclear shape through interaction with filamins. Plays an essential role in actin cytoskeleton formation in developing cartilaginous cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM101A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFLNA
Gene Accession No. Q6ZTI6
Gene Alias 3110032G18Rik; cfm; CFM2; FAM101A; family with sequence similarity 101 member A; family with sequence similarity 101, member A; filamin-interacting protein FAM101A; protein FAM101A; refilin A; RefilinA; refilin-A; regulator of filamin protein A; RFLNA; si:bz71m17.1; si:rp71-71m17.1; ZNF664-FAM101A; ZNF664-FAM101A readthrough
Gene Symbols RFLNA
Host Species Rabbit
Immunogen Recombinant Human FAM101A. Recombinant protein control fragment (Product #RP-106903).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144347
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.