Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM101A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988383
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8383 Supplier Bioss Supplier No. BS8229R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in the regulation of the perinuclear actin network and nuclear shape through interaction with filamins. Plays an essential role in actin cytoskeleton formation in developing cartilaginous cells.

Specifications

Antigen FAM101A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RFLNA
Gene Accession No. Q6ZTI6
Gene Alias 3110032G18Rik; cfm; Cfm2; Fam101a; family with sequence similarity 101 member A; family with sequence similarity 101, member A; filamin-interacting protein FAM101A; protein FAM101A; refilin A; RefilinA; Refilin-A; Regulator of filamin protein A; Rflna; si:bz71m17.1; si:rp71-71m17.1
Gene Symbols RFLNA
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM101A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144347
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.