Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM101B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556579 Supplier Invitrogen™ Supplier No. PA556579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNGTWRNYKA EVRFEPRHRP TRFLSTTIVY PKYPKAVYTT TLDYNCRKTL RRFLSSVELE AAELPGSDDL SDEC Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

RFLNB is a protein coding gene involved in the regulation of the perinuclear actin network and nuclear shape through interaction with filamins. Plays an essential role in the formation of cartilaginous skeletal elements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM101B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFLNB
Gene Accession No. Q8N5W9
Gene Alias 1500005K14Rik; cfm; Cfm1; Fam101b; family with sequence similarity 101 member B; family with sequence similarity 101, member B; filamin-interacting protein FAM101B; filamin-interacting protein FAM101B; refilin-B; MGC45871; protein FAM101B; refilin B; RefilinB; refilin-B; regulator of filamin protein B; RFLNB; RGD1359691
Gene Symbols RFLNB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM101B. Recombinant protein control fragment (Product #RP-93503).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359845
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.