Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM102A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | FAM102A |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FAM102A Polyclonal specifically detects FAM102A in Human samples. It is validated for Western Blot.Specifications
FAM102A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
bA203J24.7, C230093N12Rik, C9orf132, Early estrogen-induced gene 1 protein, Eeig1, EEIG1chromosome 9 open reading frame 132, family with sequence similarity 102, member A, MGC50853 | |
The immunogen for Anti-FAM102A antibody is: synthetic peptide directed towards the Middle region of Human F102A. Peptide sequence SLSDLTHRRNTSTSSSASGGLGMTVEGPEGSEREHRPPEKPPRPPRPLHL | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
399665 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title