Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM103A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559789
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559789 Supplier Invitrogen™ Supplier No. PA559789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDNRQFRGRD NRWGWPSDNR SNQWHGRSWG NNYPQHRQEP YYPQQYGHYG YNQRPPYGYY Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 82%.

FAM103A1, also named as RNMT-activating mini protein, is a 118 amino acid protein, which belongs to the RAM family. FAM103A1 as a component of mRNA cap binding complex localizes in the nucleus. FAM103A1 is required for efficient mRNA cap methylation and regulates RNMT expression by a post-transcriptional stabilizing mechanism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM103A1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM103A1
Gene Accession No. Q9BTL3
Gene Alias 2410047I02Rik; 2610204K14Rik; C15orf18; Fam103a1; family with sequence similarity 103 member A1; family with sequence similarity 103, member A1; HsT19360; Protein FAM103A1; RAM; Ramac; RAMMET; RGD1310022; RNA guanine-7 methyltransferase activating subunit; RNA guanine-N7 methyltransferase activating subunit; RNMT-activating mini protein; RNMT-activating mRNA cap methylating subunit; RNMT-activating mRNA cap methyltransferase subunit
Gene Symbols FAM103A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM103A1. Recombinant protein control fragment (Product #RP-98125).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83640
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.