Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM103A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559799
Description
Immunogen sequence: MTDTAEAVPK FEEMFASRFT ENDKEYQEYL KRPPESPPIV EEWNSRAGGN QRNRGNRLQ Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
FAM103A1, also named as RNMT-activating mini protein, is a 118 amino acid protein, which belongs to the RAM family. FAM103A1 as a component of mRNA cap binding complex localizes in the nucleus. FAM103A1 is required for efficient mRNA cap methylation and regulates RNMT expression by a post-transcriptional stabilizing mechanism.
Specifications
FAM103A1 | |
Polyclonal | |
Unconjugated | |
FAM103A1 | |
2410047I02Rik; 2610204K14Rik; C15orf18; Fam103a1; family with sequence similarity 103 member A1; family with sequence similarity 103, member A1; HsT19360; Protein FAM103A1; RAM; Ramac; RAMMET; RGD1310022; RNA guanine-7 methyltransferase activating subunit; RNA guanine-N7 methyltransferase activating subunit; RNMT-activating mini protein; RNMT-activating mRNA cap methylating subunit; RNMT-activating mRNA cap methyltransferase subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83640 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BTL3 | |
FAM103A1 | |
Recombinant protein corresponding to Human FAM103A1. Recombinant protein control fragment (Product #RP-98126). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction