Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM104B Antibody (OTI1D10), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272313FR
Description
FAM104B Monoclonal specifically detects FAM104B in Human samples. It is validated for Western Blot.Specifications
FAM104B | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
FAM104B | |
Full length recombinant protein of human FAM104B (NP_612371) produced in E.coli. | |
0.1 mL | |
90736.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1D10 | |
Western Blot | |
chromosome X open reading frame 44, CXorf44, family with sequence similarity 104, member B, FLJ18375, FLJ20434, hypothetical protein LOC90736 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction