Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM105A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543349
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543349 Supplier Invitrogen™ Supplier No. PA543349
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HKLKWWIGYL QRKFKRNLSV EAEVDLLSYC AREWKGETPR NKLMRKAYEE Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

FAM105A belongs to the FAM105 family. The exact function of FAM105A remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM105A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM105A
Gene Accession No. Q9NUU6
Gene Alias 9830126M18; FAM105A; family with sequence similarity 105 member A; family with sequence similarity 105, member A; inactive ubiquitin thioesterase FAM105A; Inactive ubiquitin thioesterase OTULINL; inactive ubiquitin thioesterase OTULINL; inactive ubiquitin thioesterase FAM105A; LOW QUALITY PROTEIN: inactive ubiquitin thioesterase FAM105A; NET20; OTU deubiquitinase with linear linkage specificity like; OTULINL; protein FAM105A; Protein FAM105A-like protein
Gene Symbols FAM105A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FAM105A (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54491
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.