Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FAM105B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562309
Description
Immunogen sequence: DEIEKEKELL IHERGASEPR LSVAPEMDIM DYCKKEWRGN TQKATCMKMG YEEVSQKFTS IRRVRGDNYC ALRATLFQAM SQAVGLPPWL QDPELMLLP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 89%.
Otulin is a deubiquitinase that specifically removes linear ('Met-1'-linked) polyubiquitin chains to substrates and acts as a regulator of angiogenesis and innate immune response. Required during angiogenesis, craniofacial and neuronal development by regulating the canonical Wnt signaling together with the LUBAC complex. Acts as a negative regulator of NF-kappa-B by regulating the activity of the LUBAC complex. OTULIN function is mainly restricted to homeostasis of the LUBAC complex: acts by removing 'Met-1'-linked autoubiquitination of the LUBAC complex, thereby preventing inactivation of the LUBAC complex. Acts as a key negative regulator of inflammation by restricting spontaneous inflammation and maintaining immune homeostasis. In myeloid cell, required to prevent unwarranted secretion of cytokines leading to inflammation and autoimmunity by restricting linear polyubiquitin formation. Plays a role in innate immune response by restricting linear polyubiquitin formation on LUBAC complex in response to NOD2 stimulation, probably to limit NOD2-dependent proinflammatory signaling.
Specifications
FAM105B | |
Polyclonal | |
Unconjugated | |
OTULIN | |
C79097; deubiquitinating enzyme otulin; FAM105B; family with sequence similarity 105, member B; GUM; gumby; m3Sapc; m7-1Sapc; OTU deubiquitinase with linear linkage specificity; OTU domain-containing deubiquitinase with linear linkage specificity; Otulin; protein FAM105B; ubiquitin thioesterase Gumby; Ubiquitin thioesterase otulin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90268 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BN8 | |
OTULIN | |
Recombinant protein corresponding to Human FAM105B. Recombinant protein control fragment (Product #RP-100274). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction