Learn More
Invitrogen™ FAM105B Polyclonal Antibody

Description
FAM105B Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | FAM105B |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | OTULIN |
| Gene Accession No. | Q3UCV8, Q96BN8 |
| Gene Alias | C79097; deubiquitinating enzyme otulin; FAM105B; family with sequence similarity 105, member B; GUM; gumby; m3Sapc; m7-1Sapc; OTU deubiquitinase with linear linkage specificity; OTU domain-containing deubiquitinase with linear linkage specificity; Otulin; protein FAM105B; ubiquitin thioesterase Gumby; Ubiquitin thioesterase otulin |
| Gene Symbols | OTULIN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.