Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM107A Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297556MFV610
Description
FAM107A Polyclonal antibody specifically detects FAM107A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FAM107A | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
downregulated in renal cell carcinoma, DRR1family with sequence similarity 107 member A transcript, family with sequence similarity 107, member A, FLJ30158, FLJ45473, Protein TU3A, TU3ADown-regulated in renal cell carcinoma 1 | |
Produced in rabbits immunized with E. coli-derived Human FAM107A fragment. | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication | |
11170 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction