Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM107A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563200 Supplier Invitrogen™ Supplier No. PA563200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NQLIKKKKEE LEAKRLQCPF EQELLRRQQR LNQLEKPPEK EEDHAPEFIK VRENLRRIAT LTSEER Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.

FAM107A (Family With Sequence Similarity 107 Member A) is a Protein Coding gene. Diseases associated with FAM107A include Neuroblastoma and Brain Cancer. An important paralog of this gene isFAM107B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM107A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM107A
Gene Accession No. O95990
Gene Alias Actin-associated protein FAM107A; actin-associated protein FAM107A; protein FAM107A; down-regulated in renal cell carcinoma 1; DRR1; FAM107A; family with sequence similarity 107 member A; family with sequence similarity 107, member A; protein FAM107A; Protein TU3A; RGD1306327; Tu3a
Gene Symbols FAM107A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM107A. Recombinant protein control fragment (Product #RP-91874).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11170
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.