Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM107A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595777
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595777 Supplier Invitrogen™ Supplier No. PA595777
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MYSEIQRERA DIGGLMARPE YREWNPELIK PKKLLNPVKA SRSHQELHRE LLMNHRRGLG VDSKPELQRV LEHRRRNQLI KKKKEELEAK RLQCPFEQEL LRRQQRLNQL EKPPEKEEDH APEFIKVREN LRRIATLTSE EREL; Positive Samples: Mouse brain, Rat brain; Cellular Location: Nucleus.

FAM107A (Family With Sequence Similarity 107 Member A) is a Protein Coding gene. Diseases associated with FAM107A include Neuroblastoma and Brain Cancer. An important paralog of this gene isFAM107B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM107A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FAM107A
Gene Accession No. Q78TU8
Gene Alias Actin-associated protein FAM107A; actin-associated protein FAM107A; protein FAM107A; down-regulated in renal cell carcinoma 1; DRR1; FAM107A; family with sequence similarity 107 member A; family with sequence similarity 107, member A; protein FAM107A; Protein TU3A; RGD1306327; Tu3a
Gene Symbols FAM107A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human FAM107A (NP_0091081).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268709, 361018
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.