Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM107A Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FAM107A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FAM107A Polyclonal specifically detects FAM107A in Human, Mouse samples. It is validated for Western Blot.Specifications
FAM107A | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
11170 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
downregulated in renal cell carcinoma, DRR1family with sequence similarity 107 member A transcript, family with sequence similarity 107, member A, FLJ30158, FLJ45473, Protein TU3A, TU3ADown-regulated in renal cell carcinoma 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human FAM107A (NP_009108.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title