Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM107B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | FAM107B |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
FAM107B Polyclonal specifically detects FAM107B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
FAM107B | |
Polyclonal | |
Rabbit | |
Human | |
83641 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ASFNQSGVADTHSTVRVQPVAKAGRQPRHPSAEGAPEKRQDSSTHAERNGSANRNSSHRTAAQPAETPEDVPGSLDDG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
C10orf45, chromosome 10 open reading frame 45, family with sequence similarity 107, member B, FLJ45505, hypothetical protein LOC83641, MGC11034, MGC90261 | |
FAM107B | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title