Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM107B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584394 Supplier Invitrogen™ Supplier No. PA584394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASFNQSGVAD THSTVRVQPV AKAGRQPRHP SAEGAPEKRQ DSSTHAERNG SANRNSSHRT AAQPAETPED VPGSLDDG Highest antigen sequence indentity to the following orthologs: Mouse - 27%, Rat - 28%.

FAM107B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM107B
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM107B
Gene Accession No. Q9H098
Gene Alias 3110001A13Rik; AA589595; AA959793; AI132312; axoneme-associated protein mst101(2)-like; C10orf45; Fam107b; fam107b protein; family with sequence similarity 107 member B; family with sequence similarity 107, member B; FLJ45505; HITS; hypothetical protein LOC386641; hypothetical protein MGC11034-like protein; MGC11034; MGC137524 protein; novel DUF1151 domain containing protein; protein FAM107B; wu:fc52f02; zgc:56216
Gene Symbols FAM107B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM107B. Recombinant protein control fragment (Product #RP-105114).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83641
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.