Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM108A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560304 Supplier Invitrogen™ Supplier No. PA560304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDTIEVFPTK SARGNRVSCM YVRCVPGARY Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Hydrolyzes fatty acids from S-acylated cysteine residues in proteins (PubMed:26701913). Has depalmitoylating activity towards NRAS (PubMed:26701913). Has depalmitoylating activity towards DLG4/PSD95 (PubMed:26701913). May have depalmitoylating activity towards MAP6. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM108A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD17A
Gene Accession No. Q96GS6
Gene Alias 1700013O15Rik; Abhd17a; abhydrolase domain containing 17A; abhydrolase domain containing 17A, depalmitoylase; abhydrolase domain-containing protein 17A; abhydrolase domain-containing protein FAM108A; abhydrolase domain-containing protein FAM108A1; alpha/beta hydrolase domain-containing protein 17A; BC005632; C19orf27; D10Bwg1364e; FAM108A; FAM108A1; family with sequence similarity 108, member A; family with sequence similarity 108, member A1; mFLJ00358 protein; MGC138013 protein; protein ABHD17A; RGD1359682; upf0227
Gene Symbols ABHD17A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM108A1. Recombinant protein control fragment (Product #RP-98940).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.