Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM108A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983321
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3321 Supplier Bioss Supplier No. BS14691R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen FAM108A1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ABHD17A
Gene Accession No. Q96GS6
Gene Alias 1700013O15Rik; ABHD17A; abhydrolase domain containing 17A; Abhydrolase domain-containing protein 17A; abhydrolase domain-containing protein FAM108A; abhydrolase domain-containing protein FAM108A1; Alpha/beta hydrolase domain-containing protein 17A; BC005632; BOS_8059; C19orf27; D10Bwg1364e; Fam108a; FAM108A1; family with sequence similarity 108, member A; family with sequence similarity 108, member A1; mFLJ00358 protein; protein ABHD17A; RGD1359682; upf0227
Gene Symbols ABHD17A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM108A1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81926
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.