Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM110A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567037
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567037 Supplier Invitrogen™ Supplier No. PA567037
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPVHTLSPGAP SAPALPCRLR TRVPGYLLRG PADGGARKPS AVE Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

FAM110A is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytoplasm; microtubule organizing center; spindle pole.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM110A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM110A
Gene Accession No. Q9BQ89
Gene Alias 1700008J10Rik; 5430432M24Rik; AI747080; bA371L19.3; C20orf55; chromosome 20 open reading frame 55; F10; Fam110a; family with sequence similarity 110 member A; family with sequence similarity 110, member A; protein FAM110A; RGD1308904; sb:cb450; serine threonine rich protein; si:dkey-95o9.3
Gene Symbols FAM110A
Host Species Rabbit
Immunogen Recombinant Human FAM110A. Recombinant protein control fragment (Product #RP-105466).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83541
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.